Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCDC12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001006353.1 | NP_001006353.1 | 5 - 6 | 394 | 95 | |
XM_025147149.1 | XP_025002917.1 | 5 - 6 | 350 | 95 | ||
XM_025147150.1 | XP_025002918.1 | 4 - 5 | 387 | 95 | ||
GenBank mRNA | AJ720250.1 | - | 5 - 6 | 394 | 95 |
Set Membership: |
Amplicon length between 86 and 100 Gallus gallus Probe spans exons |