Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BIRC5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001012318.1 | NP_001012318.1 | 1 - 2 | 147 | 123 | |
NM_001012319.1 | NP_001012319.1 | 1 - 2 | 147 | 123 | ||
GenBank mRNA | AF322051.1 | - | 1 - 2 | 131 | 123 | |
AF377323.1 | - | 1 - 2 | 123 | 123 | ||
BU135446.1 | - | 1 - 2 | 110 | 123 | ||
BU355446.1 | - | 1 - 2 | 140 | 123 | ||
CN227820.1 | - | 1 - 2 | 147 | 123 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Gallus gallus Probe spans exons |