Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RNF4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001012889.1 | NP_001012907.1 | 3 - 4 | 385 | 91 | |
XM_015285895.2 | XP_015141381.1 | 2 - 3 | 195 | 91 | ||
XM_015285896.2 | XP_015141382.1 | 2 - 3 | 467 | 91 | ||
GenBank mRNA | AJ719403.1 | - | 3 - 4 | 385 | 91 | |
BX930107.2 | - | 3 - 4 | 363 | 91 | ||
BX930358.2 | - | 3 - 4 | 374 | 91 |
Set Membership: |
Amplicon length between 86 and 100 Gallus gallus Probe spans exons |