Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PDCD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001031064.1 | NP_001026235.1 | 2 - 3 | 547 | 145 | |
GenBank mRNA | DQ906045.1 | - | 2 - 3 | 547 | 145 |
Set Membership: |
Amplicon length greater than or equal to 101 Gallus gallus Probe spans exons |