Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FBXO34 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001031213.1 | NP_001026384.1 | 3 - 4 | 397 | 92 | |
XM_015287511.2 | XP_015142997.1 | 5 - 6 | 579 | 92 | ||
XM_015287512.2 | XP_015142998.1 | 4 - 5 | 485 | 92 | ||
XM_015287514.2 | XP_015143000.1 | 4 - 5 | 496 | 92 | ||
XM_015287515.2 | XP_015143001.1 | 3 - 4 | 402 | 92 | ||
XM_015287516.2 | XP_015143002.1 | 3 - 4 | 406 | 92 | ||
XM_015287517.2 | XP_015143003.1 | 2 - 3 | 344 | 92 | ||
XM_025150812.1 | XP_025006580.1 | 4 - 5 | 587 | 92 | ||
GenBank mRNA | AJ720913.1 | - | 3 - 4 | 397 | 92 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Gallus gallus Probe spans exons |