Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM53A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_204388.2 | NP_989719.2 | 3 - 4 | 345 | 87 | |
XM_015285869.1 | XP_015141355.1 | 3 - 4 | 412 | 87 | ||
XM_015285871.2 | XP_015141357.1 | 3 - 4 | 342 | 87 | ||
GenBank mRNA | AF396666.1 | - | 2 - 3 | 132 | 87 | |
AJ720049.1 | - | 3 - 4 | 345 | 87 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Gallus gallus Probe spans exons |