Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMOD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_205027.1 | NP_990358.1 | 4 - 5 | 489 | 65 | |
XM_015280589.2 | XP_015136075.1 | 5 - 6 | 551 | 65 | ||
XM_015280591.2 | XP_015136077.1 | 5 - 6 | 1433 | 65 | ||
XM_025144671.1 | XP_025000439.1 | 4 - 5 | 1281 | 65 | ||
GenBank mRNA | AB052132.1 | - | 4 - 5 | 440 | 65 | |
L36678.1 | - | 4 - 5 | 489 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Gallus gallus Probe spans exons |