Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DGCR6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_205040.1 | NP_990371.1 | 3 - 4 | 612 | 85 | |
GenBank mRNA | AF048985.1 | - | 3 - 4 | 612 | 85 |
Set Membership: |
Amplicon length between 71 and 85 Gallus gallus Probe spans exons |