Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IBSP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_205162.1 | NP_990493.1 | 1 - 2 | 104 | 111 | |
GenBank mRNA | U10577.1 | - | 1 - 2 | 104 | 111 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Gallus gallus Probe spans exons |