Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CKB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_205310.1 | NP_990641.1 | 1 - 2 | 238 | 80 | |
GenBank mRNA | X03509.1 | - | 238 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Gallus gallus Probe spans exons |