Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CRABP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001030539.1 | NP_001025710.1 | 3 - 4 | 495 | 137 | |
GenBank mRNA | AB124570.1 | - | 3 - 4 | 495 | 137 | |
EU334504.1 | - | 3 - 4 | 403 | 137 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Gallus gallus Probe spans exons |