Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SHISA5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001030591.2 | NP_001025762.1 | 3 - 4 | 355 | 112 | |
XM_015293043.2 | XP_015148529.1 | 3 - 4 | 3140 | 112 | ||
GenBank mRNA | AJ720705.1 | - | 3 - 4 | 356 | 112 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Gallus gallus Probe spans exons |