Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CLTB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001030609.2 | NP_001025780.2 | 4 - 5 | 522 | 74 | |
GenBank mRNA | AJ720113.1 | - | 4 - 5 | 507 | 74 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Gallus gallus Probe spans exons |