Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SAR1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001030621.1 | NP_001025792.1 | 5 - 6 | 442 | 118 | |
XM_015294016.2 | XP_015149502.1 | 5 - 6 | 600 | 118 | ||
XM_025154854.1 | XP_025010622.1 | 5 - 6 | 386 | 118 | ||
XM_025154855.1 | XP_025010623.1 | 5 - 6 | 393 | 118 | ||
GenBank mRNA | AJ720124.1 | - | 5 - 6 | 442 | 118 |
Set Membership: |
Amplicon length greater than or equal to 101 Gallus gallus Probe spans exons |