Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: THRAP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001305665.1 | NP_001292594.1 | 7 - 8 | 2365 | 69 | |
XM_015297658.2 | XP_015153144.1 | 9 - 10 | 2599 | 69 | ||
XM_015297659.1 | XP_015153145.1 | 8 - 9 | 2390 | 69 | ||
GenBank mRNA | AJ719380.1 | - | 10 - 11 | 2614 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Gallus gallus Probe spans exons |