Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ADI1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001031089.1 | NP_001026260.1 | 3 - 4 | 454 | 71 | |
GenBank mRNA | AJ719891.1 | - | 3 - 4 | 454 | 71 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Gallus gallus Probe spans exons |