Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UFSP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001031136.1 | NP_001026307.1 | 9 - 10 | 1141 | 82 | |
XM_015276225.2 | XP_015131711.1 | 9 - 10 | 1163 | 82 | ||
XM_015276226.2 | XP_015131712.1 | 9 - 10 | 1160 | 82 | ||
GenBank mRNA | AJ720831.1 | - | 9 - 10 | 1141 | 82 | |
BX933253.1 | - | 2 - 3 | 265 | 82 |
Set Membership: |
Amplicon length between 71 and 85 Gallus gallus Probe spans exons |