Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RIC8A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001031174.1 | NP_001026345.1 | 7 - 8 | 1251 | 70 | |
XM_015286199.2 | XP_015141685.1 | 7 - 8 | 1331 | 70 | ||
XM_025150649.1 | XP_025006417.1 | 7 - 8 | 1304 | 70 | ||
GenBank mRNA | AJ719857.1 | - | 7 - 8 | 1251 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Gallus gallus Probe spans exons |