Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NDUFA10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001031247.1 | NP_001026418.1 | 6 - 7 | 793 | 85 | |
XM_015289173.2 | XP_015144659.1 | 6 - 7 | 825 | 85 | ||
XM_015289174.2 | XP_015144660.1 | 6 - 7 | 791 | 85 | ||
GenBank mRNA | AJ720729.1 | - | 6 - 7 | 793 | 85 |
Set Membership: |
Amplicon length between 71 and 85 Gallus gallus Probe spans exons |