Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DTWD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001031442.1 | NP_001026613.1 | 4 - 5 | 535 | 112 | |
GenBank mRNA | AJ719497.1 | - | 4 - 5 | 535 | 112 |
Set Membership: |
Amplicon length greater than or equal to 101 Gallus gallus Probe spans exons |