Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DCLRE1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001031500.3 | NP_001026671.3 | 919 | 81 | ||
GenBank mRNA | AY376897.1 | - | 919 | 81 | ||
BX950430.1 | - | 537 | 81 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Gallus gallus |