Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NTPCR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001031523.1 | NP_001026694.1 | 1 - 2 | 56 | 76 | |
GenBank mRNA | AJ719769.1 | - | 1 - 2 | 56 | 76 | |
KT153608.1 | - | 1 - 2 | 33 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Gallus gallus Probe spans exons |