Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LHX3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001030335.1 | NP_001025506.1 | 1 - 2 | 70 | 90 | |
XM_025156012.1 | XP_025011780.1 | 1 - 2 | 137 | 90 | ||
XM_025156013.1 | XP_025011781.1 | 1 - 2 | 137 | 90 | ||
GenBank mRNA | AF461699.1 | - | 1 - 2 | 100 | 90 | |
L35570.1 | - | 1 - 2 | 70 | 90 |
Set Membership: |
Amplicon length between 86 and 100 Recommended Assay Gallus gallus Probe spans exons |