Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LHX3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001030335.1 | NP_001025506.1 | 5 - 6 | 771 | 76 | |
XM_025156012.1 | XP_025011780.1 | 4 - 5 | 991 | 76 | ||
GenBank mRNA | L35570.1 | - | 5 - 6 | 771 | 76 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Gallus gallus Probe spans exons |