Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NAMPTP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001030728.1 | NP_001025899.1 | 1 - 2 | 113 | 78 | |
XM_015275716.2 | XP_015131202.1 | 1 - 2 | 366 | 78 | ||
GenBank mRNA | AJ719830.1 | - | 1 - 2 | 153 | 78 | |
AY946242.1 | - | 1 - 2 | 113 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Gallus gallus Probe spans exons |