Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NAMPTP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001030728.1 | NP_001025899.1 | 5 - 6 | 669 | 90 | |
XM_015275716.2 | XP_015131202.1 | 5 - 6 | 922 | 90 | ||
GenBank mRNA | AY946242.1 | - | 5 - 6 | 669 | 90 |
Set Membership: |
Amplicon length between 86 and 100 Gallus gallus Probe spans exons |