Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PWP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001030761.2 | NP_001025932.2 | 9 - 10 | 957 | 119 | |
GenBank mRNA | AJ720939.1 | - | 9 - 10 | 964 | 119 | |
AJ727221.1 | - | 4 - 5 | 369 | 119 |
Set Membership: |
Amplicon length greater than or equal to 101 Gallus gallus Probe spans exons |