Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CDV3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001030990.1 | NP_001026161.1 | 3 - 4 | 629 | 69 | |
XM_015281501.1 | XP_015136987.1 | 3 - 4 | 626 | 69 | ||
XM_025147217.1 | XP_025002985.1 | 3 - 4 | 626 | 69 | ||
GenBank mRNA | AJ719755.1 | - | 3 - 4 | 629 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Gallus gallus Probe spans exons |