Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TSHR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001033850.1 | NP_001029022.1 | 3 - 4 | 352 | 135 | |
NM_001193588.1 | NP_001180517.1 | 5 - 6 | 499 | 135 | ||
NM_001193589.1 | NP_001180518.1 | 4 - 5 | 424 | 135 | ||
XM_025150837.1 | XP_025006605.1 | 5 - 6 | 682 | 135 | ||
GenBank mRNA | AB234613.1 | - | 5 - 6 | 499 | 135 | |
AB234614.1 | - | 4 - 5 | 424 | 135 | ||
AB234615.1 | - | 3 - 4 | 352 | 135 |
Set Membership: |
Amplicon length greater than or equal to 101 Gallus gallus Probe spans exons |