Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TAPBP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034816.3 | NP_001029988.2 | 2 - 3 | 266 | 134 | |
NM_001206611.2 | NP_001193540.1 | 2 - 3 | 266 | 134 | ||
GenBank mRNA | AJ005072.1 | - | 1 - 2 | 102 | 134 | |
AM403065.1 | - | 2 - 3 | 236 | 134 | ||
AM403066.1 | - | 2 - 3 | 236 | 134 | ||
AM403067.1 | - | 2 - 3 | 236 | 134 | ||
AM403068.1 | - | 2 - 3 | 236 | 134 | ||
AM403069.1 | - | 2 - 3 | 236 | 134 | ||
AM403070.1 | - | 2 - 3 | 236 | 134 | ||
AM403071.1 | - | 2 - 3 | 236 | 134 | ||
AM403072.1 | - | 2 - 3 | 236 | 134 | ||
AM403073.1 | - | 2 - 3 | 236 | 134 |
Set Membership: |
Amplicon length greater than or equal to 101 Recommended Assay Gallus gallus Probe spans exons |