Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WNT7B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001037274.1 | NP_001032351.1 | 3 - 4 | 642 | 71 | |
NM_001171593.1 | NP_001165064.1 | 3 - 4 | 909 | 71 | ||
GenBank mRNA | AY753290.1 | - | 3 - 4 | 642 | 71 | |
AY753291.1 | - | 3 - 4 | 909 | 71 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Gallus gallus Probe spans exons |