Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KCTD7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001039269.1 | NP_001034358.1 | 3 - 4 | 645 | 84 | |
GenBank mRNA | AJ720387.1 | - | 3 - 4 | 645 | 84 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Gallus gallus Probe spans exons |