Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SRPRA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001039277.1 | NP_001034366.1 | 4 - 5 | 575 | 93 | |
GenBank mRNA | AJ851760.1 | - | 4 - 5 | 575 | 93 |
Set Membership: |
Amplicon length between 86 and 100 Gallus gallus Probe spans exons |