Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HNRNPAB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_205328.5 | NP_990659.1 | 5 - 6 | 861 | 81 | |
XM_015293750.2 | XP_015149236.1 | 6 - 7 | 4007 | 81 | ||
XM_015293751.2 | XP_015149237.1 | 6 - 7 | 4007 | 81 | ||
XM_015293752.2 | XP_015149238.1 | 6 - 7 | 4007 | 81 | ||
GenBank mRNA | CR352392.1 | - | 6 - 7 | 759 | 81 | |
U14942.1 | - | 5 - 6 | 808 | 81 | ||
X80340.1 | - | 865 | 81 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Gallus gallus Probe spans exons |