Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARHGAP11B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001006479.2 | NP_001006479.2 | 3 - 4 | 353 | 76 | |
XM_015287342.2 | XP_015142828.1 | 3 - 4 | 400 | 76 | ||
XM_015287343.2 | XP_015142829.1 | 3 - 4 | 254 | 76 | ||
XM_025150725.1 | XP_025006493.1 | 5 - 6 | 5427 | 76 | ||
GenBank mRNA | AY313784.1 | - | 3 - 4 | 353 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Gallus gallus Probe spans exons |