Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COLEC11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001044665.1 | NP_001038130.1 | 4 - 5 | 338 | 88 | |
XM_015284842.2 | XP_015140328.1 | 5 - 6 | 9049 | 88 | ||
XM_015284843.2 | XP_015140329.1 | 5 - 6 | 561 | 88 | ||
XM_015284844.2 | XP_015140330.1 | 4 - 5 | 453 | 88 | ||
XM_025148811.1 | XP_025004579.1 | 4 - 5 | 252 | 88 | ||
GenBank mRNA | DQ129669.1 | - | 4 - 5 | 338 | 88 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Gallus gallus Probe spans exons |