Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DACT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001044692.1 | NP_001038157.1 | 3 - 4 | 670 | 80 | |
GenBank mRNA | DQ066927.1 | - | 3 - 4 | 670 | 80 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Gallus gallus Probe spans exons |