Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DLX1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001045842.2 | NP_001039307.2 | 1 - 2 | 453 | 88 | |
GenBank mRNA | AY640309.1 | - | 1 - 2 | 453 | 88 | |
CR386811.1 | - | 1 - 2 | 454 | 88 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Gallus gallus Probe spans exons |