Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NTHL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001079575.1 | NP_001073043.1 | 3 - 4 | 462 | 75 | |
GenBank mRNA | AB284187.1 | - | 3 - 4 | 462 | 75 | |
BX930531.1 | - | 3 - 4 | 475 | 75 | ||
CR353294.1 | - | 3 - 4 | 635 | 75 | ||
CR390660.1 | - | 2 - 3 | 310 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Gallus gallus Probe spans exons |