Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NLGN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081502.1 | NP_001074971.1 | 4 - 5 | 759 | 92 | |
XM_015291652.2 | XP_015147138.1 | 5 - 6 | 1685 | 92 | ||
XM_015291653.2 | XP_015147139.1 | 5 - 6 | 1731 | 92 | ||
XM_015291654.2 | XP_015147140.1 | 5 - 6 | 1227 | 92 | ||
XM_015291655.2 | XP_015147141.1 | 5 - 6 | 1687 | 92 | ||
XM_015291656.2 | XP_015147142.1 | 4 - 5 | 1624 | 92 | ||
XM_015291658.2 | XP_015147144.1 | 4 - 5 | 1625 | 92 | ||
XM_015291659.2 | XP_015147145.1 | 3 - 4 | 1564 | 92 | ||
XM_015291660.2 | XP_015147146.1 | 3 - 4 | 1565 | 92 | ||
GenBank mRNA | EF199811.1 | - | 4 - 5 | 759 | 92 | |
GQ489204.1 | - | 4 - 5 | 759 | 92 | ||
GU080337.1 | - | 4 - 5 | 759 | 92 | ||
GU080338.1 | - | 3 - 4 | 699 | 92 | ||
GU080339.1 | - | 3 - 4 | 699 | 92 | ||
GU080340.1 | - | 2 - 3 | 639 | 92 | ||
GU574580.1 | - | 4 - 5 | 759 | 92 |
Set Membership: |
Amplicon length between 86 and 100 Recommended Assay Gallus gallus Probe spans exons |