Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TNFSF11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001083361.1 | NP_001076830.1 | 2 - 3 | 474 | 71 | |
GenBank mRNA | EF379383.1 | - | 2 - 3 | 474 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Gallus gallus Probe spans exons |