Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GLI2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001271901.1 | NP_001258830.1 | 4 - 5 | 538 | 76 | |
NM_001271902.1 | NP_001258831.1 | 4 - 5 | 435 | 76 | ||
XM_015289786.2 | XP_015145272.1 | 4 - 5 | 753 | 76 | ||
XM_015289787.2 | XP_015145273.1 | 4 - 5 | 754 | 76 | ||
XM_025152142.1 | XP_025007910.1 | 5 - 6 | 685 | 76 | ||
GenBank mRNA | AF022818.1 | - | 4 - 5 | 442 | 76 | |
AM262745.1 | - | 4 - 5 | 501 | 76 | ||
AM262746.1 | - | 4 - 5 | 538 | 76 | ||
AM262747.1 | - | 2 - 3 | 337 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Gallus gallus Probe spans exons |