Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TYR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_204160.1 | NP_989491.1 | 1 - 2 | 927 | 77 | |
XM_015279812.2 | XP_015135298.1 | 1 - 2 | 1004 | 77 | ||
GenBank mRNA | AB023291.1 | - | 1 - 2 | 868 | 77 | |
D88349.1 | - | 1 - 2 | 927 | 77 | ||
MG672516.1 | - | 1 - 2 | 821 | 77 | ||
MG672517.1 | - | 1 - 2 | 821 | 77 | ||
S81675.1 | - | 1 - 2 | 863 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Gallus gallus Probe spans exons |