Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UNG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_204189.1 | NP_989520.1 | 5 - 6 | 581 | 65 | |
GenBank mRNA | AF525946.1 | - | 5 - 6 | 581 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Gallus gallus Probe spans exons |