Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: POU1F1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_204319.1 | NP_989650.1 | 1 - 2 | 268 | 82 | |
XM_015295651.2 | XP_015151137.1 | 2 - 3 | 2388 | 82 | ||
XM_015295656.2 | XP_015151142.1 | 2 - 3 | 2304 | 82 | ||
GenBank mRNA | AF029892.1 | - | 1 - 2 | 268 | 82 | |
AJ222692.2 | - | 1 - 2 | 184 | 82 | ||
AJ236855.1 | - | 2 - 3 | 208 | 82 | ||
AJ236856.1 | - | 2 - 3 | 292 | 82 |
Set Membership: |
Amplicon length between 71 and 85 Gallus gallus Probe spans exons |