Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TNFSF10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015291627.2 | XP_015147113.1 | 5 - 6 | 843 | 64 | |
XM_015291628.2 | XP_015147114.1 | 4 - 5 | 449 | 64 | ||
XM_015291629.2 | XP_015147115.1 | 5 - 6 | 797 | 64 | ||
GenBank mRNA | AB114678.1 | - | 4 - 5 | 455 | 64 | |
AM066390.1 | - | 3 - 4 | 428 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Gallus gallus Probe spans exons |