Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RHOQ | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_204461.1 | NP_989792.1 | 3 - 4 | 400 | 148 | |
GenBank mRNA | AF372468.1 | - | 3 - 4 | 400 | 148 |
Set Membership: |
Amplicon length greater than or equal to 101 Gallus gallus Probe spans exons |