Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC35A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_204513.1 | NP_989844.1 | 6 - 7 | 808 | 107 | |
XM_015284597.2 | XP_015140083.1 | 6 - 7 | 907 | 107 | ||
GenBank mRNA | AJ535486.1 | - | 6 - 7 | 808 | 107 | |
BX929414.2 | - | 6 - 7 | 882 | 107 | ||
BX930442.2 | - | 6 - 7 | 817 | 107 |
Set Membership: |
Amplicon length greater than or equal to 101 Gallus gallus Probe spans exons |