Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ITM2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_205085.2 | NP_990416.2 | 5 - 6 | 861 | 83 | |
XM_015275287.2 | XP_015130773.1 | 5 - 6 | 848 | 83 | ||
GenBank mRNA | AF017985.1 | - | 5 - 6 | 723 | 83 | |
CN223991.1 | - | 3 - 4 | 458 | 83 | ||
DR420019.1 | - | 823 | 83 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Gallus gallus Probe spans exons |