Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HMGN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_205106.2 | NP_990437.2 | 4 - 5 | 461 | 65 | |
GenBank mRNA | AJ441772.1 | - | 4 - 5 | 461 | 65 | |
CV859154.1 | - | 3 - 4 | 221 | 65 | ||
M20817.1 | - | 4 - 5 | 319 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Gallus gallus Probe spans exons |